5-Nitro-2,4,6-trichloropyrimidine ≥98%

Supplier: Ambeed
Warning

A761500-5G A761500-250MG A761500-1G
77552-334EA 154 USD
77552-334 77747-503 77747-502
5-Nitro-2,4,6-trichloropyrimidine ≥98%
5-Nitro-2,4,6-trichloropyrimidine

Precaución: for Research Use Only

Formula: C₄Cl₃N₃O₂
MDL Number: MFCD00091516
CAS Number: 4359-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR