tert-Butyl 2-(2-(2-(2-bromoethoxy)ethoxy)ethoxy)acetate ≥95%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG3-CH2-Boc

A760318-1G A760318-25G A760318-100MG A760318-250MG A760318-5G
77552-328EA 256 USD
77552-328 77783-972 77783-970 77783-971 77783-973
tert-Butyl 2-(2-(2-(2-bromoethoxy)ethoxy)ethoxy)acetate ≥95%
tert-Butyl 2-(2-(2-(2-bromoethoxy)ethoxy)ethoxy)acetate

Precaución: for Research Use Only

Formula: C₁₂H₂₃BrO₅
MDL Number: MFCD30723231
CAS Number: 2100306-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR