1,3-Propanediol ≥99%

Supplier: Ambeed
Warning

A757959-100G A757959-25G A757959-1000G
77552-322EA 17 USD
77552-322 77552-324 77573-444
1,3-Propanediol ≥99%
1,3-Propanediol

Precaución: for Research Use Only

Formula: HO(CH₂)₃OH
MDL Number: MFCD00002949
CAS Number: 504-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR