Cobalt(II) iodide 98%

Supplier: Ambeed
Warning

A757509-1G A757509-25G A757509-5G
77552-318EA 8.24 USD
77552-318 77552-320 77569-448
Cobalt(II) iodide 98%
Cobalt(II) iodide

Precaución: For research use only.

Formula: CoI₂
MDL Number: MFCD00016030
CAS Number: 15238-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR