Allicin ≥97% (mixture of isomers)

Supplier: Ambeed
Danger

A757336-5G A757336-25G
77552-316EA 31 USD
77552-316 77602-944
Allicin ≥97% (mixture of isomers)
Allicin

Precaución: for Research Use Only

Formula: C₆H₁₀OS₂
MDL Number: MFCD00468100
CAS Number: 539-86-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR