tert-Butyl 2-(2-bromoethoxy)acetate ≥95%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG1-CH2-Boc

A755250-1G A755250-5G A755250-100MG A755250-250MG
77552-312EA 57.68 USD
77552-312 77573-434 77598-978 77622-788
tert-Butyl 2-(2-bromoethoxy)acetate ≥95%
tert-Butyl 2-(2-bromoethoxy)acetate

Precaución: for Research Use Only

Formula: C₈H₁₅BrO₃
MW: 239.1069 g/mol
MDL Number: MFCD28122952
CAS Number: 157759-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR