Pent-4-yn-1-amine ≥95%

Supplier: Ambeed
Danger

A753371-1G A753371-250MG A753371-5G
77552-302EA 257.5 USD
77552-302 77552-304 77573-422
Pent-4-yn-1-amine ≥95%
Pent-4-yn-1-amine

Precaución: for Research Use Only

Formula: C₅H₉N
MW: 83.131 g/mol
MDL Number: MFCD09923611
CAS Number: 15252-44-5
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR