(6Z,9Z,28Z,31Z)-Heptatriaconta-6,9,28,31-tetraen-19-yl 4-(dimethylamino)butanoate 98%

Supplier: Ambeed
Warning

Synonyms: D-Lin-MC3-DMA

A746322-1G A746322-250MG A746322-50MG A746322-100MG A746322-5G A746322-10MG
77552-280EA 1126.82 USD
77552-280 77552-282 77552-284 77569-412 77569-414 77573-384
(6Z,9Z,28Z,31Z)-Heptatriaconta-6,9,28,31-tetraen-19-yl 4-(dimethylamino)butanoate 98%
(6Z,9Z,28Z,31Z)-Heptatriaconta-6,9,28,31-tetraen-19-yl 4-(dimethylamino)butanoate

Precaución: For research use only.

Formula: C₄₃H₇₉NO₂
MW: 642.09 g/mol
MDL Number: MFCD31657426
CAS Number: 1224606-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR