3-Chloro-3-methyl-1-butyne 97%

Supplier: Ambeed
Danger

A743075-25G A743075-10G
77552-278EA 26 USD
77552-278 77569-404
3-Chloro-3-methyl-1-butyne 97%
3-Chloro-3-methyl-1-butyne

Precaución: For research use only.

Formula: C₅H₇Cl
MDL Number: MFCD00190221
CAS Number: 1111-97-3
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR