3-Bromo-4-methyl-4H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A726064-1G A726064-250MG
77552-248EA 563.41 USD
77552-248 77569-380
3-Bromo-4-methyl-4H-1,2,4-triazole 98%
3-Bromo-4-methyl-4H-1,2,4-triazole

Precaución: For research use only.

Formula: C₃H₄BrN₃
MW: 161.99 g/mol
MDL Number: MFCD10696422
CAS Number: 16681-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR