3-(4-Aminophenyl)piperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A725837-1G A725837-5G A725837-250MG
77552-246EA 670 USD
77552-246 77569-378 77573-336
3-(4-Aminophenyl)piperidine-2,6-dione 97%
3-(4-Aminophenyl)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD24561931
CAS Number: 92137-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR