(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl 2-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate

A719980-100MG A719980-250MG
77552-226EA 32.96 USD
77552-226 77552-228
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester ≥97%
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₅
MW: 259.302 g/mol
MDL Number: MFCD02259744
CAS Number: 321744-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR