Ethyl iododifluoroacetate ≥98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2,2-difluoro-2-iodoacetate, Ethyl difluoro(iodo)acetate

A719120-5G A719120-1G
77552-216EA 37 USD
77552-216 77602-842
Ethyl iododifluoroacetate ≥98%
Ethyl iododifluoroacetate

Precaución: for Research Use Only

Formula: C₄H₅F₂IO₂
MDL Number: MFCD00798135
CAS Number: 7648-30-8
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR