Fmoc-L-Glu(2-phenylisopropyloxy)-OH ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Glu(2-phenylisopropyl ester)-OH

A717153-1G A717153-5G A717153-250MG A717153-100MG
77552-196EA 360.5 USD
77552-196 77552-198 77614-202 77684-637
Fmoc-L-Glu(2-phenylisopropyloxy)-OH ≥97%
Fmoc-L-Glu(2-phenylisopropyloxy)-OH

Precaución: For research use only.

Formula: C₂₉H₂₉NO₆
MW: 487.54 g/mol
MDL Number: MFCD00672713
CAS Number: 200616-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR