6-Bromohexan-1-amine hydrobromide ≥97%

Supplier: Ambeed
Warning

A707100-1G A707100-100MG
77552-158EA 49 USD
77552-158 77747-411
6-Bromohexan-1-amine hydrobromide ≥97%
6-Bromohexan-1-amine hydrobromide

Precaución: for Research Use Only

Formula: C₆H₁₅Br₂N
MDL Number: MFCD17170307
CAS Number: 14502-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR