Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) ≥98%

Supplier: Ambeed
Danger

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), Ni(TMHD)2

A706424-1G A706424-5G A706424-25G
77552-154EA 46.35 USD
77552-154 77552-156 77627-444
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) ≥98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II)

Precaución: for Research Use Only

Formula: C₂₂H₃₈NiO₄
MW: 425.23 g/mol
MDL Number: MFCD00192348
CAS Number: 14481-08-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR