Benzyl isocyanide ≥98%

Supplier: Ambeed
Danger

A705689-1G A705689-5G A705689-25G
77552-148EA 23 USD
77552-148 77552-150 77878-543
Benzyl isocyanide ≥98%
Benzyl isocyanide

Precaución: for Research Use Only

Formula: C₈H₇N
MW: 117.15 g/mol
MDL Number: MFCD00000004
CAS Number: 10340-91-7
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR