Nitrosium tetrafluoroborate ≥95%

Supplier: Ambeed
Danger

Synonyms: Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate

A702196-1G A702196-5G A702196-25G
77552-130EA 13.39 USD
77552-130 77552-132 77602-788
Nitrosium tetrafluoroborate ≥95%
Nitrosium tetrafluoroborate

Precaución: for Research Use Only

Formula: BF₄NO
MW: 116.81 g/mol
MDL Number: MFCD00011433
CAS Number: 14635-75-7
UN: 3260
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR