Methyl 5-(bromomethyl)-2-nitrobenzoate 95%

Supplier: Ambeed
Danger

A701578-5G A701578-25G A701578-1G
77552-124EA 153 USD
77552-124 77569-336 77617-900
Methyl 5-(bromomethyl)-2-nitrobenzoate 95%
Methyl 5-(bromomethyl)-2-nitrobenzoate

Precaución: For research use only.

Formula: C₉H₈BrNO₄
MDL Number: MFCD20528191
CAS Number: 88071-91-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR