D(+)-α-Acetobromoglucose ≥98% stabilised

Supplier: Ambeed
Warning

Synonyms: 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose

Stabilization: Stabilised with calcium carbonate 1-2%

A686595-25G A686595-5G A686595-100G A686595-500G
77552-084EA 16 USD
77552-084 77552-086 77569-306 77569-308
D(+)-α-Acetobromoglucose ≥98% stabilised
D(+)-α-Acetobromoglucose

Precaución: For research use only.

Formula: C₁₄H₁₉BrO₉
MDL Number: MFCD00063254
CAS Number: 572-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR