3,3'-Dihydroxy-[1,1'-biphenyl]-4,4'-dicarboxylic acid ≥97%

Supplier: Ambeed
Warning

A684329-1G A684329-250MG A684329-5G
77552-082EA 126.69 USD
77552-082 77598-704 77617-848
3,3'-Dihydroxy-[1,1'-biphenyl]-4,4'-dicarboxylic acid ≥97%
3,3'-Dihydroxy-[1,1'-biphenyl]-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₆
MW: 274.2256 g/mol
MDL Number: MFCD24387008
CAS Number: 861533-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR