(R)-2-Methylazetidine hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
A681923-1G
A681923-100MG
A681923-250MG
77552-072EA
219
USD
77552-072
77891-342
77891-343
(R)-2-Methylazetidine hydrochloride ≥97%
(R)-2-Methylazetidine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₀ClN |
MDL Number:
MFCD29920003 CAS Number: 791614-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...