Ethyl 4-(prop-2-ynylamino)benzoate ≥95%

Supplier: Ambeed
Warning

A679244-5G A679244-1G A679244-250MG
77552-066EA 207 USD
77552-066 77617-826 77617-828
Ethyl 4-(prop-2-ynylamino)benzoate ≥95%
Ethyl 4-(prop-2-ynylamino)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD19596318
CAS Number: 101248-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR