Benzyl 3-(aminomethyl)azetidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A679061-5G A679061-250MG A679061-1G
77552-064EA 163 USD
77552-064 77614-096 77673-850
Benzyl 3-(aminomethyl)azetidine-1-carboxylate ≥98%
Benzyl 3-(aminomethyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MW: 220.26 g/mol
MDL Number: MFCD07772065
CAS Number: 1016731-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR