Pelargonaldehyde ≥98%

Supplier: Ambeed
Warning

A672210-25G A672210-100G A672210-500G
77552-048EA 10 USD
77552-048 77573-164 77669-830
Pelargonaldehyde ≥98%
Pelargonaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₈O
MDL Number: MFCD00007030
CAS Number: 124-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR