2,4-Dichloropteridine ≥98%

Supplier: Ambeed
Warning

A669983-250MG A669983-100MG
77552-040EA 255 USD
77552-040 77886-588
2,4-Dichloropteridine ≥98%
2,4-Dichloropteridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₄
MDL Number: MFCD09743914
CAS Number: 98138-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR