(2-Oxo-1,2-dihydropyridin-3-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A662306-1G A662306-100MG A662306-250MG A662306-5G
77552-022EA 109 USD
77552-022 77886-570 77886-571 77886-572
(2-Oxo-1,2-dihydropyridin-3-yl)boronic acid ≥98%
(2-Oxo-1,2-dihydropyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₆BNO₃
MDL Number: MFCD12964572
CAS Number: 951655-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR