Dimethyldivinylsilane 98

Supplier: Ambeed
Danger

A658489-25G A658489-5G
77552-008EA 29 USD
77552-008 77880-384
Dimethyldivinylsilane 98
Dimethyldivinylsilane

Precaución: for Research Use Only

Formula: C₆H₁₂Si
MW: 112.25 g/mol
MDL Number: MFCD00053855
CAS Number: 10519-87-6
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR