α,α-Difluorophenylacetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: α,α-Difluorobenzeneacetic acid

A656309-5G A656309-25G
77551-998EA 59.74 USD
77551-998 77686-624
α,α-Difluorophenylacetic acid ≥98%
α,α-Difluorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD00498146
CAS Number: 360-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR