Palladium hydroxide 20% (w/w) on carbon

Supplier: Ambeed
Danger

A656225-10G A656225-25G A656225-5G A656225-1G
77551-992EA 122.57 USD
77551-992 77551-994 77551-996 77569-246
Palladium hydroxide 20% (w/w) on carbon
Palladium hydroxide

Precaución: For research use only.

Formula: H₂O₂Pd
MDL Number: MFCD00064599
CAS Number: 12135-22-7
UN: 3178
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR