Ethenesulfonyl chloride 95%

Supplier: Ambeed
Danger

A652973-1G A652973-250MG A652973-5G A652973-50MG
77551-980EA 218 USD
77551-980 77551-982 77569-236 77573-090
Ethenesulfonyl chloride 95%
Ethenesulfonyl chloride

Precaución: For research use only.

Formula: C₂H₃ClO₂S
MW: 126.562 g/mol
MDL Number: MFCD19200056
CAS Number: 6608-47-5
UN: 3265
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR