2-Bromo-5-iodoaniline ≥96%

Supplier: Ambeed
Warning

A651355-5G A651355-1G
77551-962EA 432.6 USD
77551-962 77684-539
2-Bromo-5-iodoaniline ≥96%
2-Bromo-5-iodoaniline

Precaución: For research use only.

Formula: C₆H₅BrIN
MDL Number: MFCD16038675
CAS Number: 111721-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR