3,3-Difluoropropan-1-amine hydrochloride ≥98%

Supplier: Ambeed
Danger

A639405-100MG A639405-250MG A639405-5G
77551-946EA 154 USD
77551-946 77617-694 77749-307
3,3-Difluoropropan-1-amine hydrochloride ≥98%
3,3-Difluoropropan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈ClF₂N
MDL Number: MFCD12024563
CAS Number: 1010097-89-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR