5-Chloro-2'-deoxyuridine ≥98%

Supplier: Ambeed
Warning

A639135-1G A639135-5G A639135-100MG A639135-250MG
77551-944EA 131 USD
77551-944 77747-316 77747-314 77747-315
5-Chloro-2'-deoxyuridine ≥98%
5-Chloro-2'-deoxyuridine

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂O₅
MDL Number: MFCD00006531
CAS Number: 50-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR