Potassium 2,2,2-trifluoroethane-2-trifluoroborate ≥95%

Supplier: Ambeed
Warning

A638543-1G A638543-250MG
77551-942EA 143 USD
77551-942 77893-278
Potassium 2,2,2-trifluoroethane-2-trifluoroborate ≥95%
Potassium 2,2,2-trifluoroethane-2-trifluoroborate

Precaución: for Research Use Only

Formula: C₂H₂BF₆·K
MDL Number: MFCD27981361
CAS Number: 1510778-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR