Indium(III) chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: Indium Trichloride Chloride, Indium trichloride

A635854-10G A635854-25G A635854-5G A635854-1G
77551-924EA 21.63 USD
77551-924 77551-926 77551-928 77573-060
Indium(III) chloride ≥98%
Indium(III) chloride

Precaución: for Research Use Only

Formula: Cl₃In
MW: 221.18 g/mol
MDL Number: MFCD00011058
CAS Number: 10025-82-8
UN: 3260
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR