(S)-a-(Fmoc-N-methylamino)benzenebutanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-Me-HoPhe-OH

A633020-5G A633020-1G
77551-910EA 388.31 USD
77551-910 77594-952
(S)-a-(Fmoc-N-methylamino)benzenebutanoic acid ≥97%
(S)-a-(Fmoc-N-methylamino)benzenebutanoic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₄
MW: 415.4810 g/mol
MDL Number: MFCD06796749
CAS Number: 1065076-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR