(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-([1,1'-biphenyl]-3-yl)propanoic acid ≥95%

Supplier: Ambeed
Warning

A631989-1G A631989-100MG A631989-250MG A631989-5G
77551-906EA 242 USD
77551-906 77783-528 77783-529 77783-530
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-([1,1'-biphenyl]-3-yl)propanoic acid ≥95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-([1,1'-biphenyl]-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₃₀H₂₅NO₄
MDL Number: MFCD07372099
CAS Number: 1260616-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR