N2-Isobutyryl-2'-O-methylguanosine ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(9-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-3H-purin-2-yl)isobutyramide

A625719-5G A625719-250MG
77551-872EA 266.77 USD
77551-872 77686-569
N2-Isobutyryl-2'-O-methylguanosine ≥98%
N2-Isobutyryl-2'-O-methylguanosine

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₅O₆
MW: 367.35 g/mol
MDL Number: MFCD15145297
CAS Number: 63264-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR