5-Amino-3-bromo-1-methyl-1H-pyrazole ≥98%

Supplier: Ambeed
Warning

A625057-1G A625057-250MG A625057-100MG
77551-870EA 220 USD
77551-870 77594-918 77673-690
5-Amino-3-bromo-1-methyl-1H-pyrazole ≥98%
5-Amino-3-bromo-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₆BrN₃
MDL Number: MFCD18800831
CAS Number: 1357471-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR