tert-Butyl 6-hydroxy-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥98%

Supplier: Ambeed
Warning

A623906-1G A623906-250MG
77551-866EA 800.31 USD
77551-866 77573-026
tert-Butyl 6-hydroxy-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥98%
tert-Butyl 6-hydroxy-3-azabicyclo[3.1.0]hexane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.246 g/mol
MDL Number: MFCD31567464
CAS Number: 1540700-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR