Hexacarbonylmolybdenum ≥99%

Supplier: Ambeed
Danger

Synonyms: Molybdenum carbonyl, Hexacarbonylmolybdenum(0), Molybdenumhexacarbonyl, Molybdenum hexacarbonyl

A620890-1G A620890-25G
77551-862EA 41 USD
77551-862 77569-188
Hexacarbonylmolybdenum ≥99%
Hexacarbonylmolybdenum

Precaución: For research use only.

Formula: C₆MoO₆
MDL Number: MFCD00003466
CAS Number: 13939-06-5
UN: 3466
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR