4-Chloro-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidine ≥97%

Supplier: Ambeed
Warning

A618768-1G A618768-5G
77551-858EA 85.49 USD
77551-858 77573-014
4-Chloro-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidine ≥97%
4-Chloro-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄S
MW: 200.64 g/mol
MDL Number: MFCD09702036
CAS Number: 85426-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR