Tributylphosphine oxide ≥97%

Supplier: Ambeed
Danger

Synonyms: Tri-n-butylphosphineoxide, Oseltamivir impurity H

A614959-5G A614959-25G
77551-842EA 8 USD
77551-842 77669-640
Tributylphosphine oxide ≥97%
Tributylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₂H₂₇OP
MW: 218.32 g/mol
Boiling Pt: 348
MDL Number: MFCD00002082
CAS Number: 814-29-9
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR