2,2-Dimethoxy-2-phenylacetophenone ≥98%

Supplier: Ambeed
Danger

A614214-25G A614214-100G A614214-1000G
77551-836EA 5 USD
77551-836 77686-549 77900-742
2,2-Dimethoxy-2-phenylacetophenone ≥98%
2,2-Dimethoxy-2-phenylacetophenone

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MDL Number: MFCD00008475
CAS Number: 24650-42-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR