3-Phenylpropylamine ≥98%

Supplier: Ambeed
Danger

Synonyms: 3-Phenyl-1-propylamine

A612688-25G A612688-5G
77551-832EA 38.11 USD
77551-832 77551-834
3-Phenylpropylamine ≥98%
3-Phenylpropylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00008224
CAS Number: 2038-57-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR