Fmoc-Arg(Me,Pbf)-OH ≥98%

Supplier: Ambeed
Warning

A604296-1G A604296-100MG A604296-250MG
77551-818EA 172 USD
77551-818 77882-418 77882-419
Fmoc-Arg(Me,Pbf)-OH ≥98%
Fmoc-Arg(Me,Pbf)-OH

Precaución: for Research Use Only

Formula: C₃₅H₄₂N₄O₇S
MDL Number: MFCD08064312
CAS Number: 1135616-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR