(SP-4-3)-[[2′-(Diphenylphosphino)[1,1′-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium ≥98%

Supplier: Ambeed
Warning

A598714-1G A598714-5G A598714-100MG
77551-804EA 352 USD
77551-804 77572-930 77747-235
(SP-4-3)-[[2′-(Diphenylphosphino)[1,1′-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium ≥98%
(SP-4-3)-[[2′-(Diphenylphosphino)[1,1′-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium

Precaución: for Research Use Only

Formula: C₅₈H₄₇NO₃P₂PdS
MDL Number: MFCD31707673
CAS Number: 1599466-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR