1-Hexanol ≥99%

Supplier: Ambeed
Danger

Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane

A592872-25G A592872-100G
77551-788EA 9.27 USD
77551-788 77594-804
1-Hexanol ≥99%
1-Hexanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₄CH₂OH
MW: 102.18 g/mol
MDL Number: MFCD00002982
CAS Number: 111-27-3
UN: 2282
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR