trans-tert-Butyl 4-aminocyclohexanecarboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-butyl trans-4-aminocyclohexanecarboxylate

A589999-1G A589999-250MG A589999-5G A589999-100MG
77551-774EA 70.04 USD
77551-774 77617-476 77617-478 77747-225
trans-tert-Butyl 4-aminocyclohexanecarboxylate ≥98%
trans-tert-Butyl 4-aminocyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₂
MW: 199.29 g/mol
MDL Number: MFCD30475674
CAS Number: 1022159-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR